This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02479907_g1
See other ZTF-9 GE Assays ›
Gene Symbol:
ztf-9
Gene Name:
Zinc finger putative Transcription Factor family
Gene Aliases:
CELE_ZK287.6
Chromosome Location:
Chr.V: 9691045 - 9693316 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02479907_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ztf-9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_073096.5 NP_505497.1 2 - 3 226 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

nucleic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0