This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02475984_g1
See other TTN-1 GE Assays ›
Gene Symbol:
ttn-1
Gene Name:
protein-coding
Gene Aliases:
CELE_W06H8.8
Chromosome Location:
Chr.V: 6120909 - 6202632 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02475984_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ttn-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029029.5 NP_001024200.1 9 - 10 3785 112
NM_001359765.3 NP_001346723.1 2408 112
NM_001359766.3 NP_001346716.1 2408 112
NM_001359767.3 NP_001346722.1 3686 112
NM_001359768.3 NP_001346715.1 3686 112
NM_001359777.3 NP_001346724.1 8 - 9 2495 112
NM_001373034.1 NP_001360072.1 3779 112
NM_001380656.1 NP_001367914.1 2408 112
NM_001380657.1 NP_001367912.1 2501 112
NM_001380658.2 NP_001367913.1 3686 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
myosin binding
metal ion binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0