This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02471076_m1
See other MADD-2 GE Assays ›
Gene Symbol:
madd-2
Gene Name:
protein-coding
Gene Aliases:
CELE_C39F7.2
Chromosome Location:
Chr.V: 1223888 - 1235455 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02471076_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: madd-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129405.4 NP_001122877.1 11 - 12 2178 107
NM_001313132.3 NP_001300061.1 1 - 2 54 107
NM_070994.7 NP_503395.1 12 - 13 2210 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

axon midline choice point recognition
protein ubiquitination
cell projection assembly
neuron projection development
regulation of protein localization
dorsal/ventral axon guidance
ubiquitin-protein transferase activity
receptor binding
protein binding
zinc ion binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0