This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02469404_m1
See other EGL-8 GE Assays ›
Gene Symbol:
egl-8
Gene Name:
protein-coding
Gene Aliases:
CELE_B0348.4
Chromosome Location:
Chr.V: 20818 - 55451 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02469404_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: egl-8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026175.4 NP_001021346.1 5 - 6 954 64
NM_001026176.3 NP_001021347.1 5 - 6 954 64
NM_001026177.6 NP_001021348.1 5 - 6 960 64
NM_001313085.3 NP_001300014.1 4 772 64
NM_001313087.3 NP_001300016.1 5 - 6 954 64
NM_001313089.4 NP_001300018.1 6 961 64
NM_001313091.3 NP_001300020.1 6 960 64
NM_001313093.3 NP_001300022.1 6 960 64
NM_001313095.4 NP_001300024.1 4 823 64
NM_001313097.3 NP_001300026.1 3 688 64
NM_001313099.3 NP_001300028.1 3 688 64
NM_001313101.3 NP_001300030.1 3 688 64
NM_001313103.4 NP_001300032.1 4 - 5 870 64
NM_001313105.3 NP_001300034.1 3 688 64
NM_001372980.2 NP_001360389.1 6 960 64
NM_001372988.2 NP_001360397.1 5 876 64
NM_001380576.2 NP_001368552.1 5 - 6 772 64
NM_001380577.1 NP_001368553.1 5 - 6 960 64
NM_001380578.2 NP_001368554.1 5 - 6 958 64
NM_001380579.1 NP_001368555.1 5 - 6 960 64
NM_001380580.2 NP_001368556.1 4 - 5 872 64
NM_001380581.2 NP_001368557.1 4 - 5 876 64
NM_001380582.2 NP_001368558.1 4 - 5 858 64
NM_001380583.1 NP_001368559.1 4 - 5 851 64
NM_001380584.1 NP_001368560.1 4 - 5 688 64
GenBank mRNA AF179426.1 - 5 - 6 954 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
chemotaxis
signal transduction
dopamine receptor signaling pathway
positive regulation of acetylcholine secretion, neurotransmission
lipid catabolic process
protein localization to synapse
intracellular signal transduction
regulation of locomotion
positive regulation of locomotion
regulation of pharyngeal pumping
thermotaxis
regulation of oviposition
phosphatidylinositol-mediated signaling
positive regulation of axon regeneration
phosphatidylinositol phospholipase C activity
calcium ion binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0