This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02469402_g1
See other EGL-8 GE Assays ›
Gene Symbol:
egl-8
Gene Name:
protein-coding
Gene Aliases:
CELE_B0348.4
Chromosome Location:
Chr.V: 20818 - 55451 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02469402_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: egl-8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026175.4 NP_001021346.1 3 - 4 768 75
NM_001026176.3 NP_001021347.1 3 - 4 768 75
NM_001026177.6 NP_001021348.1 3 - 4 774 75
NM_001313085.3 NP_001300014.1 2 586 75
NM_001313087.3 NP_001300016.1 3 - 4 768 75
NM_001313089.4 NP_001300018.1 4 775 75
NM_001313091.3 NP_001300020.1 4 774 75
NM_001313093.3 NP_001300022.1 4 774 75
NM_001372980.2 NP_001360389.1 4 774 75
NM_001380576.2 NP_001368552.1 3 - 4 586 75
NM_001380577.1 NP_001368553.1 3 - 4 774 75
NM_001380578.2 NP_001368554.1 3 - 4 772 75
NM_001380579.1 NP_001368555.1 3 - 4 774 75
GenBank mRNA AF179426.1 - 3 - 4 768 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
chemotaxis
signal transduction
dopamine receptor signaling pathway
positive regulation of acetylcholine secretion, neurotransmission
lipid catabolic process
protein localization to synapse
intracellular signal transduction
regulation of locomotion
positive regulation of locomotion
regulation of pharyngeal pumping
thermotaxis
regulation of oviposition
phosphatidylinositol-mediated signaling
positive regulation of axon regeneration
phosphatidylinositol phospholipase C activity
calcium ion binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0