This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02468930_g1
See other ITSN-1 GE Assays ›
Gene Symbol:
itsn-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y116A8C.36
Chromosome Location:
Chr.IV: 17119682 - 17126269 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02468930_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: itsn-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268998.3 NP_001255927.1 4 - 5 1424 82
NM_001268999.3 NP_001255928.1 2 - 3 642 82
NM_001269000.3 NP_001255929.1 1 - 2 63 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
endosomal transport
clathrin-dependent synaptic vesicle endocytosis
positive regulation of synaptic vesicle endocytosis
protein binding
binding, bridging

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0