This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02467915_g1
See other EPS-8 GE Assays ›
Gene Symbol:
eps-8
Gene Name:
protein-coding
Gene Aliases:
CELE_Y57G11C.24
Chromosome Location:
Chr.IV: 14879484 - 14900653 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02467915_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eps-8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001028350.6 NP_001023521.2 13 - 14 2282 71
NM_001028351.5 NP_001023522.2 10 - 11 1744 71
NM_001047582.5 NP_001041047.1 15 - 16 2574 71
NM_001306998.2 NP_001293927.1 16 - 17 2648 71
NM_001306999.3 NP_001293928.1 15 - 16 2408 71
NM_001307000.3 NP_001293929.1 14 - 15 2357 71
NM_001307001.3 NP_001293930.1 11 - 12 1817 71
NM_001307002.3 NP_001293931.1 3 - 4 284 71
NM_001372963.2 NP_001360610.1 16 2576 71
NM_001372964.2 NP_001360611.1 15 2501 71
NM_070402.6 NP_502803.3 14 - 15 2333 71
GenBank mRNA AY611484.1 - 15 - 16 2498 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nematode larval development
Rho protein signal transduction
regulation of Rho protein signal transduction
intermediate filament organization
maintenance of protein location
digestive tract morphogenesis
embryonic morphogenesis
epidermis morphogenesis
barbed-end actin filament capping
actin filament bundle assembly
actin binding
protein binding
cytoskeletal protein binding
Rac guanyl-nucleotide exchange factor activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0