This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02467678_m1
See other XDH-1 GE Assays ›
Gene Symbol:
xdh-1
Gene Name:
Xanthine DeHydrogenase homolog
Gene Aliases:
CELE_F55B11.1
Chromosome Location:
Chr.IV: 14401114 - 14412894 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02467678_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: xdh-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_070346.4 NP_502747.1 8 - 9 1754 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

xanthine catabolic process
electron transport chain
oxidation-reduction process
xanthine dehydrogenase activity
xanthine oxidase activity
iron ion binding
electron carrier activity
oxidoreductase activity
metal ion binding
flavin adenine dinucleotide binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header