This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02467251_g1
See other ISCU-1 GE Assays ›
Gene Symbol:
iscu-1
Gene Name:
NifU_N domain-containing protein
Gene Aliases:
CELE_Y45F10D.4
Chromosome Location:
Chr.IV: 13783962 - 13784680 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02467251_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: iscu-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_070257.4 NP_502658.1 1 - 2 105 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Endogenous Controls Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cellular iron ion homeostasis
iron-sulfur cluster assembly
iron ion binding
ferrous iron binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0