This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02466729_g1
See other MSH-5 GE Assays ›
Gene Symbol:
msh-5
Gene Name:
protein-coding
Gene Aliases:
CELE_F09E8.3
Chromosome Location:
Chr.IV: 13155233 - 13164054 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02466729_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: msh-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001380506.1 NP_001368472.1 9 - 10 2454 102
NM_070130.8 NP_502531.1 19 - 20 3792 102
GenBank mRNA AF271389.1 - 19 - 20 3792 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

meiotic mismatch repair
mismatch repair
reciprocal meiotic recombination
homologous chromosome segregation
chiasma assembly
meiotic cell cycle
nucleotide binding
DNA binding
protein binding
ATP binding
DNA-dependent ATPase activity
mismatched DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0