This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02465925_g1
See other DPY-26 GE Assays ›
Gene Symbol:
dpy-26
Gene Name:
Condensin complex subunit dpy-26
Gene Aliases:
CELE_C25G4.5
Chromosome Location:
Chr.IV: 12444507 - 12451335 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02465925_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dpy-26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_069977.8 NP_502378.1 4 - 5 2127 82
GenBank mRNA U43562.1 - 4 - 5 2082 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

mitotic sister chromatid segregation
cell cycle
chromosome condensation
dosage compensation by hypoactivation of X chromosome
meiotic chromosome segregation
meiotic sister chromatid segregation
cell division
meiotic cell cycle

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0