This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02465835_g1
See other CCR-4 GE Assays ›
Gene Symbol:
ccr-4
Gene Name:
protein-coding
Gene Aliases:
CELE_ZC518.3
Chromosome Location:
Chr.IV: 12361542 - 12371852 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02465835_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ccr-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001028436.4 NP_001023607.1 6 - 7 1253 76
NM_001028437.4 NP_001023608.1 6 - 7 1257 76
NM_001136405.2 NP_001129877.1 3 - 4 1604 76
NM_001307061.3 NP_001293990.1 2 - 3 502 76
NM_069954.6 NP_502355.2 6 - 7 1230 76
GenBank mRNA AY043268.1 - 6 - 7 1120 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA poly(A) tail shortening
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
exonuclease activity
poly(A)-specific ribonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0