This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02464554_g1
See other Y73B6BL.14 GE Assays ›
Gene Symbol:
Y73B6BL.14
Gene Name:
DNA_ligase_A_N domain-containing protein
Gene Aliases:
CELE_Y73B6BL.14
Chromosome Location:
Chr.IV: 6341432 - 6342976 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02464554_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Y73B6BL.14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_068569.4 NP_500970.2 1 - 2 111 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA ligation
lagging strand elongation
DNA repair
DNA recombination
DNA ligation involved in DNA repair
Okazaki fragment processing involved in mitotic DNA replication
DNA binding
DNA ligase (ATP) activity
ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0