This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02463912_m1
See other UNC-5 GE Assays ›
Gene Symbol:
unc-5
Gene Name:
protein-coding
Gene Aliases:
CELE_B0273.4
Chromosome Location:
Chr.IV: 5495664 - 5528338 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02463912_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306820.3 NP_001293749.1 5 - 6 684 93
NM_001306822.3 NP_001293751.1 1 - 2 99 93
NM_001383098.1 4 - 5 605 93
NM_068421.6 NP_500822.2 4 - 5 600 93
NM_068422.6 NP_500823.1 4 - 5 602 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
multicellular organism development
ventral cord development
motor neuron axon guidance
mesodermal cell migration
establishment or maintenance of actin cytoskeleton polarity
negative regulation of microtubule polymerization
dorsal/ventral axon guidance
gonad morphogenesis
netrin-activated signaling pathway
positive regulation of locomotion
nematode male tail tip morphogenesis
positive regulation of axon extension
chemorepulsion of axon
commissural neuron axon guidance
positive regulation of anterior/posterior axon guidance
regulation of dorsal/ventral axon guidance
netrin receptor activity
protein tyrosine kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0