This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02463792_g1
See other HDA-6 GE Assays ›
Gene Symbol:
hda-6
Gene Name:
protein-coding
Gene Aliases:
CELE_F41H10.6
Chromosome Location:
Chr.IV: 5356543 - 5364306 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02463792_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hda-6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129308.4 NP_001122780.1 3 - 4 660 71
NM_068386.4 NP_500787.1 3 - 4 677 71
NM_068387.4 NP_500788.1 3 - 4 677 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
protein deacetylation
histone deacetylation
histone H3 deacetylation
histone deacetylase activity
zinc ion binding
hydrolase activity
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0