This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02463303_g1
See other TAT-2 GE Assays ›
Gene Symbol:
tat-2
Gene Name:
protein-coding
Gene Aliases:
CELE_H06H21.10
Chromosome Location:
Chr.IV: 4827853 - 4846893 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02463303_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tat-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001028081.3 NP_001023252.2 14 - 15 2462 89
NM_001307515.2 NP_001294444.1 14 - 15 2462 89
NM_001307516.1 NP_001294445.1 15 - 16 2567 89
NM_001307517.3 NP_001294446.1 15 - 16 2567 89
NM_001307518.3 NP_001294447.1 15 - 16 2567 89
NM_001380211.1 NP_001367920.1 14 - 15 2462 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

Golgi organization
phospholipid transport
sterol metabolic process
phospholipid translocation
nucleotide binding
magnesium ion binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0