This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02461149_m1
See other CPSF-1 GE Assays ›
Gene Symbol:
cpsf-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y76B12C.7
Chromosome Location:
Chr.IV: 2009038 - 2032996 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02461149_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cpsf-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001380077.1 NP_001368311.1 9 - 10 2361 70
NM_067756.6 NP_500157.2 12 - 13 2938 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0