This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02460569_m1
See other PLX-1 GE Assays ›
Gene Symbol:
plx-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y55F3AL.1
Chromosome Location:
Chr.IV: 959521 - 984395 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02460569_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plx-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307625.3 NP_001294554.1 22 - 23 5368 66
NM_001307626.3 NP_001294555.1 19 - 20 4801 66
NM_001307627.3 NP_001294556.1 19 - 20 4768 66
NM_001307628.3 NP_001294557.1 10 - 11 3022 66
NM_001307629.3 NP_001294558.1 10 - 11 3055 66
NM_067617.6 NP_500018.3 22 - 23 5335 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell adhesion
signal transduction
small GTPase mediated signal transduction
regulation of cell shape
epidermis development
regulation of cell migration
regulation of GTPase activity
nematode male tail tip morphogenesis
positive regulation of axonogenesis
regulation of cellular component movement
semaphorin-plexin signaling pathway
nematode male tail mating organ morphogenesis
semaphorin-plexin signaling pathway involved in axon guidance
protein binding
semaphorin receptor activity
Rac GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0