This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02458667_g1
See other DIS-3 GE Assays ›
Gene Symbol:
dis-3
Gene Name:
Probable exosome complex exonuclease RRP44
Gene Aliases:
CELE_C04G2.6
Chromosome Location:
Chr.IV: 10097196 - 10106523 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02458667_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dis-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_069434.5 NP_501835.2 10 - 11 2519 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
rRNA catabolic process
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
RNA binding
nuclease activity
endonuclease activity
exonuclease activity
ribonuclease activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0