This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02458478_m1
See other GON-1 GE Assays ›
Gene Symbol:
gon-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F25H8.3
Chromosome Location:
Chr.IV: 9934206 - 9953433 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02458478_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gon-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268519.3 NP_001255448.1 6 - 7 795 63
NM_001372913.1 NP_001359558.1 9 - 10 1299 63
NR_056474.1 9 - 10 1295 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
ER to Golgi vesicle-mediated transport
integrin-mediated signaling pathway
multicellular organism development
gonad development
protein transport
vesicle-mediated transport
cell migration
extracellular matrix organization
muscle organ morphogenesis
negative regulation of presynapse assembly
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0