This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02457294_g1
See other PDZD-8 GE Assays ›
Gene Symbol:
pdzd-8
Gene Name:
protein-coding
Gene Aliases:
CELE_C53B4.4
Chromosome Location:
Chr.IV: 8975526 - 8984579 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02457294_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pdzd-8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027922.6 NP_001023093.1 5 - 6 801 90
NM_001129292.5 NP_001122764.2 5 - 6 912 90
NM_001307127.3 NP_001294056.1 5 - 6 801 90
NM_001307128.3 NP_001294057.1 5 - 6 801 90
NM_001372894.3 NP_001360498.1 5 - 6 912 90
NM_001372895.3 NP_001360499.1 5 801 90
NM_001372896.2 NP_001360495.1 5 - 6 912 90
NM_001372897.2 NP_001360496.1 5 - 6 912 90
NM_001372898.2 NP_001360497.1 5 - 6 912 90
NM_069157.5 NP_501558.2 5 - 6 905 90
NM_069158.4 NP_501559.2 5 - 6 906 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
intracellular signal transduction
mitochondrial calcium ion homeostasis
mitochondrion-ER tethering
lipid binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header