This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02457289_g1
See other PDZD-8 GE Assays ›
Gene Symbol:
pdzd-8
Gene Name:
protein-coding
Gene Aliases:
CELE_C53B4.4
Chromosome Location:
Chr.IV: 8975526 - 8984579 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02457289_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pdzd-8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027922.6 NP_001023093.1 16 - 17 3731 68
NM_001307128.3 NP_001294057.1 16 - 17 3737 68
NM_001372894.3 NP_001360498.1 17 - 18 4034 68
NM_001372895.3 NP_001360499.1 17 3917 68
NM_069157.5 NP_501558.2 17 - 18 4018 68
NM_069158.4 NP_501559.2 17 - 18 4013 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
intracellular signal transduction
mitochondrial calcium ion homeostasis
mitochondrion-ER tethering
lipid binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0