This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02455873_g1
See other ITR-1 GE Assays ›
Gene Symbol:
itr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F33D4.2
Chromosome Location:
Chr.IV: 7675174 - 7696930 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02455873_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: itr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027999.4 NP_001023170.1 21 - 22 5562 104
NM_001028000.5 NP_001023171.1 19 - 20 5394 104
NM_001028002.4 NP_001023173.1 20 - 21 5490 104
NM_001268376.4 NP_001255305.1 21 - 22 5487 104
GenBank mRNA AF168688.1 - 20 - 21 5490 104
AJ243179.2 - 20 - 21 5512 104
AJ243180.2 - 21 - 22 5585 104
AJ243181.2 - 20 - 21 5505 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
ion transport
calcium ion transport
muscle contraction
positive regulation of cytosolic calcium ion concentration
multicellular organism development
rhythmic behavior
programmed cell death
regulation of cell migration
defecation
sperm ejaculation
regulation of pharyngeal pumping
inositol phosphate-mediated signaling
inositol lipid-mediated signaling
embryonic morphogenesis
multicellular organismal reproductive process
hermaphrodite genitalia morphogenesis
release of sequestered calcium ion into cytosol
regulation of filopodium assembly
transmembrane transport
male mating behavior
calcium ion transmembrane transport
positive regulation of pharyngeal pumping
ion channel activity
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium channel activity
calcium ion binding
protein binding
myosin binding
phosphatidylinositol binding
inositol 1,4,5 trisphosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0