This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02454575_m1
See other TAF-9 GE Assays ›
Gene Symbol:
taf-9
Gene Name:
TAF (TBP-associated transcription factor) family
Gene Aliases:
CELE_T12D8.7
Chromosome Location:
Chr.III: 13624276 - 13626007 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02454575_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: taf-9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_067413.4 NP_499814.1 2 - 3 436 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA-templated transcription, initiation
transcription from RNA polymerase II promoter
embryo development ending in birth or egg hatching
regulation of mitotic cell cycle, embryonic
histone H3 acetylation
positive regulation of transcription from RNA polymerase II promoter
transcription coactivator activity
obsolete general RNA polymerase II transcription factor activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0