This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02454370_g1
See other F45G2.8 GE Assays ›
Gene Symbol:
F45G2.8 F45G2.7
Gene Name:
Mitochondrial import inner membrane translocase subunit tim-16
Uncharacterized protein
Gene Aliases:
CELE_F45G2.8
CELE_F45G2.7
Chromosome Location:
Chr.III: 13434861 - 13435670 on Build WBcel235
Chromosome Location:
Chr.III: 13433439 - 13435670 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02454370_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F45G2.8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_067374.6 NP_499775.1 1 - 2 94 66
Gene Symbol: F45G2.7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383025.1 1 - 2 94 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001383025.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

protein transport
protein import into mitochondrial matrix

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0