This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02452562_m1
See other PDE-12 GE Assays ›
Gene Symbol:
pde-12
Gene Name:
Endo/exonuclease/phosphatase domain-containing protein
Gene Aliases:
CELE_R17.2
Chromosome Location:
Chr.III: 10840005 - 10845020 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02452562_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pde-12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066987.5 NP_499388.1 5 - 6 1698 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
exonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0