This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02451727_g1
See other T16G12.1 GE Assays ›
Gene Symbol:
T16G12.1
Gene Name:
Putative aminopeptidase-2
Gene Aliases:
CELE_T16G12.1
Chromosome Location:
Chr.III: 10040181 - 10049857 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02451727_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: T16G12.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066829.4 NP_499230.2 19 - 20 3514 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
embryo development ending in birth or egg hatching
cell differentiation
peptide catabolic process
regulation of oviposition
oogenesis
regulation of meiotic cell cycle process involved in oocyte maturation
aminopeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
peptide binding
metal ion binding
metalloaminopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0