This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02451419_g1
See other CBP-1 GE Assays ›
Gene Symbol:
cbp-1
Gene Name:
protein-coding
Gene Aliases:
CELE_R10E11.1
Chromosome Location:
Chr.III: 9766510 - 9774990 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02451419_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cbp-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129239.3 NP_001122711.2 11 - 12 6045 82
NM_066759.5 NP_499160.2 11 - 12 6171 82
NM_066760.7 NP_499161.2 11 - 12 6132 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
regulation of transcription, DNA-templated
apoptotic process
spermatogenesis
embryo development ending in birth or egg hatching
regulation of neuron projection development
histone acetylation
internal peptidyl-lysine acetylation
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor binding
transcription cofactor activity
transcription coactivator activity
histone acetyltransferase activity
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
transcription factor binding
zinc ion binding
transferase activity
transferase activity, transferring acyl groups
chromatin DNA binding
metal ion binding
RNA polymerase II sequence-specific DNA binding transcription factor binding
peptide-lysine-N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0