This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02449614_g1
See other GRP-1 GE Assays ›
Gene Symbol:
grp-1 idi-1
Gene Name:
GTP exchange factor for ARFs 1
Nudix hydrolase domain-containing protein
Gene Aliases:
CELE_K06H7.4
CELE_K06H7.9
Chromosome Location:
Chr.III: 8085705 - 8088449 on Build WBcel235
Chromosome Location:
Chr.III: 8085699 - 8093046 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02449614_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: grp-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066363.4 NP_498764.1 3 - 4 351 57
Gene Symbol: idi-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001382931.1 3 - 4 364 57

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001382931.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of ARF protein signal transduction
isoprenoid biosynthetic process
isopentenyl diphosphate biosynthetic process
guanyl-nucleotide exchange factor activity
ARF guanyl-nucleotide exchange factor activity
isopentenyl-diphosphate delta-isomerase activity
hydrolase activity
isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0