This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02449131_g1
See other UPP-1 GE Assays ›
Gene Symbol:
upp-1
Gene Name:
Uridine and thymidine phosphorylase
Gene Aliases:
CELE_ZK783.2
Chromosome Location:
Chr.III: 7641034 - 7643101 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02449131_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: upp-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066270.7 NP_498671.2 3 - 4 815 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

uracil catabolic process
uridine catabolic process
pyrimidine nucleotide biosynthetic process
nucleoside metabolic process
nucleotide catabolic process
uracil metabolic process
UMP salvage
thymidine metabolic process
uridine metabolic process
catalytic activity
uridine phosphorylase activity
thymidine phosphorylase activity
pyrimidine-nucleoside phosphorylase activity
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0