This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02448553_g1
See other FUBL-2 GE Assays ›
Gene Symbol:
fubl-2 tofu-7
Gene Name:
protein-coding
Twenty One u-rna (21U-RNA) biogenesis Fouled Up
Gene Aliases:
CELE_ZK418.9
CELE_ZK418.8
Chromosome Location:
Chr.III: 7085196 - 7087747 on Build WBcel235
Chromosome Location:
Chr.III: 7083684 - 7086594 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02448553_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fubl-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027829.5 NP_001023000.1 5 - 6 883 64
NM_001382907.1 5 - 6 894 64
Gene Symbol: tofu-7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066146.7 NP_498547.1 1 - 2 210 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_066146.7 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
mRNA processing
regulation of gene expression
nucleic acid binding
RNA binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0