This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02447914_g1
See other T26A5.6 GE Assays ›
Gene Symbol:
T26A5.6 let-721
Gene Name:
UDENN domain-containing protein
Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial
Gene Aliases:
CELE_T26A5.6
CELE_C05D11.12
Chromosome Location:
Chr.III: 6447615 - 6450696 on Build WBcel235
Chromosome Location:
Chr.III: 6444692 - 6450698 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02447914_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: T26A5.6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066015.5 NP_498416.2 3 - 4 498 140
Gene Symbol: let-721
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001382896.1 3 - 4 500 140

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001382896.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

electron transport chain
respiratory electron transport chain
oxidation-reduction process
electron-transferring-flavoprotein dehydrogenase activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, oxidizing metal ions with flavin as acceptor
metal ion binding
ubiquinone binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0