This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02447724_g1
See other GEN-1 GE Assays ›
Gene Symbol:
gen-1
Gene Name:
GEN1 Holliday junction resolvase homolog
Gene Aliases:
CELE_T12A2.8
Chromosome Location:
Chr.III: 6245917 - 6248158 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02447724_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gen-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065960.4 NP_498361.1 8 - 9 1063 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA repair
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of nematode larval development
signal transduction involved in DNA damage checkpoint
nucleic acid phosphodiester bond hydrolysis
four-way junction DNA binding
DNA binding
catalytic activity
nuclease activity
crossover junction endodeoxyribonuclease activity
5'-flap endonuclease activity
flap endonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0