This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02447456_m1
See other NONO-1 GE Assays ›
Gene Symbol:
nono-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F25B5.7
Chromosome Location:
Chr.III: 5953526 - 5958255 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02447456_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nono-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001372604.2 NP_001359971.1 5 - 6 1105 90
NM_065894.4 NP_498295.3 5 - 6 1101 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header