This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02447308_g1
See other POLQ-1 GE Assays ›
Gene Symbol:
polq-1
Gene Name:
DNA polymerase theta
Gene Aliases:
CELE_W03A3.2
Chromosome Location:
Chr.III: 5791119 - 5803160 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02447308_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: polq-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001379729.2 NP_001367994.1 15 - 16 3528 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA-dependent DNA replication
DNA repair
base-excision repair, gap-filling
cellular response to DNA damage stimulus
cellular response to ionizing radiation
DNA biosynthetic process
nucleotide binding
nucleic acid binding
DNA binding
DNA-directed DNA polymerase activity
ATP binding
5'-3' exonuclease activity
transferase activity
nucleotidyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0