This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02447166_m1
See other RCAN-1 GE Assays ›
Gene Symbol:
rcan-1
Gene Name:
Calcipressin-like protein
Gene Aliases:
CELE_F54E7.7
Chromosome Location:
Chr.III: 5682301 - 5684098 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02447166_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rcan-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065822.7 NP_498223.2 3 - 4 569 79
GenBank mRNA AF176115.1 - 3 - 4 546 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

calcium-mediated signaling
negative regulation of phosphoprotein phosphatase activity
response to hermaphrodite contact
vulval location
thermosensory behavior
regulation of calcineurin-NFAT signaling cascade
positive regulation of nematode male tail tip morphogenesis
nucleic acid binding
protein serine/threonine phosphatase inhibitor activity
protein binding
calcium-dependent protein serine/threonine phosphatase regulator activity
protein phosphatase 2B binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0