This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02446860_g1
See other Y32H12A.8 GE Assays ›
Gene Symbol:
Y32H12A.8
Gene Name:
protein-coding
Gene Aliases:
CELE_Y32H12A.8
Chromosome Location:
Chr.III: 5385489 - 5406037 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02446860_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Y32H12A.8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001372583.2 NP_001360479.1 7 801 89
NM_001372585.2 NP_001360485.1 6 - 7 789 89
NM_001379709.1 NP_001368729.1 6 - 7 813 89
NM_001379710.2 NP_001368730.1 7 - 8 845 89
NM_065750.6 NP_498151.2 6 - 7 792 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0