This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02446631_g1
See other PRMT-5 GE Assays ›
Gene Symbol:
prmt-5
Gene Name:
Protein arginine N-methyltransferase 5
Gene Aliases:
CELE_C34E10.5
Chromosome Location:
Chr.III: 5230570 - 5234530 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02446631_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: prmt-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065711.8 NP_498112.1 2 - 3 391 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
regulation of transcription, DNA-templated
protein methylation
apoptotic process
cellular response to DNA damage stimulus
locomotory behavior
intrinsic apoptotic signaling pathway in response to DNA damage
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
methylation
histone arginine methylation
peptidyl-arginine N-methylation
negative regulation of DNA damage response, signal transduction by p53 class mediator
histone H4-R3 methylation
negative regulation of transcription, DNA-templated
response to odorant
p53 binding
methyltransferase activity
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
[myelin basic protein]-arginine N-methyltransferase activity
transferase activity
activating transcription factor binding
protein-arginine omega-N monomethyltransferase activity
protein-arginine omega-N symmetric methyltransferase activity
histone methyltransferase activity (H4-R3 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0