This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02446326_g1
See other MRPS-18C GE Assays ›
Gene Symbol:
mrps-18C fsn-1
Gene Name:
Putative 39S ribosomal protein L3, mitochondrial
F-box/SPRY domain-containing protein 1
Gene Aliases:
CELE_C26E6.6
CELE_C26E6.5
Chromosome Location:
Chr.III: 4935578 - 4937227 on Build WBcel235
Chromosome Location:
Chr.III: 4935576 - 4939457 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02446326_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mrps-18C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065644.6 NP_498045.2 3 - 4 707 91
Gene Symbol: fsn-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065645.7 NP_498046.1 3 - 4 709 91

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_065645.7 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

translation
ubiquitin-dependent protein catabolic process
determination of adult lifespan
regulation of synaptic growth at neuromuscular junction
response to ionizing radiation
negative regulation of gene expression
protein ubiquitination
negative regulation of axon extension
dauer larval development
negative regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of synaptic growth at neuromuscular junction
negative regulation of axon regeneration
positive regulation of synapse assembly
synapse assembly involved in innervation
regulation of neuromuscular synaptic transmission
positive regulation of neuromuscular synaptic transmission
positive regulation of protein localization to nucleus
structural constituent of ribosome
protease binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0