This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02445962_g1
See other MRPL-16 GE Assays ›
Gene Symbol:
mrpl-16 pgap-2
Gene Name:
Ribosomal_L16 domain-containing protein
Post-GPI attachment to proteins factor 2
Gene Aliases:
CELE_T04A8.11
CELE_T04A8.12
Chromosome Location:
Chr.III: 4704404 - 4706946 on Build WBcel235
Chromosome Location:
Chr.III: 4704406 - 4706953 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02445962_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mrpl-16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065564.7 NP_497965.2 5 - 6 1354 117
Gene Symbol: pgap-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001384079.1 6 - 7 1070 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

translation
mitochondrial translation
GPI anchor biosynthetic process
structural constituent of ribosome
rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0