This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02445035_m1
See other NLP-10 GE Assays ›
Gene Symbol:
nlp-10
Gene Name:
Neuropeptide-Like Protein
Gene Aliases:
CELE_F37A8.4
Chromosome Location:
Chr.III: 3933574 - 3935908 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02445035_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nlp-10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_065394.6 NP_497795.1 1 - 2 103 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

neuropeptide signaling pathway

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0