This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02444530_g1
See other HECW-1 GE Assays ›
Gene Symbol:
hecw-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F45H7.6
Chromosome Location:
Chr.III: 3377396 - 3389970 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02444530_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hecw-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001330981.3 NP_001317872.1 21 - 22 3691 60
NM_065296.4 NP_497697.2 18 - 19 3191 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
ubiquitin-dependent protein catabolic process
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
regulation of dendrite morphogenesis
regulation of defense response to bacterium
negative regulation of sodium ion transmembrane transporter activity
ubiquitin-protein transferase activity
transferase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0