Assay Details
Gene Symbol: mltn-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064637.7 | NP_497038.2 | 5 - 6 | 1237 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: mltn-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064637.7 | NP_497038.2 | 5 - 6 | 1237 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() |