This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02440410_g1
See other CNP-2 GE Assays ›
Gene Symbol:
cnp-2
Gene Name:
Calcineurin-interacting protein 2
Gene Aliases:
CELE_Y46G5A.10
Chromosome Location:
Chr.II: 12760797 - 12767277 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02440410_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cnp-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_064314.6 NP_496715.3 12 - 13 1630 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0