This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02440102_g1
See other DAF-22 GE Assays ›
Gene Symbol:
daf-22
Gene Name:
Non-specific lipid-transfer protein-like 2
Gene Aliases:
CELE_Y57A10C.6
Chromosome Location:
Chr.II: 12423492 - 12425694 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02440102_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: daf-22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_064238.6 NP_496639.1 3 - 4 522 66
GenBank mRNA D86473.1 - 3 - 4 544 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
acyl-CoA metabolic process
lipid transport
olfactory learning
negative regulation of lipid storage
response to pheromone
positive regulation of intracellular cholesterol transport
pheromone biosynthetic process
dauer entry
dauer exit
positive regulation of developmental growth
regulation of chemotaxis
positive regulation of nematode larval development
ascaroside biosynthetic process
catalytic activity
lipid binding
transferase activity
transferase activity, transferring acyl groups other than amino-acyl groups
propanoyl-CoA C-acyltransferase activity
propionyl-CoA C2-trimethyltridecanoyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0