This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02439247_g1
See other AARS-1 GE Assays ›
Gene Symbol:
aars-1
Gene Name:
Alanine--tRNA ligase, mitochondrial
Gene Aliases:
CELE_W02B12.6
Chromosome Location:
Chr.II: 11460205 - 11464497 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02439247_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: aars-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_064043.7 NP_496444.1 5 - 6 2112 78
NR_023953.1 5 - 6 1610 78
GenBank mRNA AF188715.1 - 5 - 6 1691 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

tRNA modification
translation
alanyl-tRNA aminoacylation
tRNA aminoacylation
aminoacyl-tRNA metabolism involved in translational fidelity
tRNA binding
nucleotide binding
aminoacyl-tRNA editing activity
RNA binding
aminoacyl-tRNA ligase activity
alanine-tRNA ligase activity
ATP binding
zinc ion binding
amino acid binding
ligase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0