This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02438921_g1
See other MAT-2 GE Assays ›
Gene Symbol:
mat-2 apc-17
Gene Name:
Anaphase-promoting complex subunit 1
Anaphase Promoting Complex; see also mat
Gene Aliases:
CELE_W10C6.1
CELE_C09H10.7
Chromosome Location:
Chr.II: 11112719 - 11121107 on Build WBcel235
Chromosome Location:
Chr.II: 11108128 - 11120776 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02438921_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mat-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063982.4 NP_496383.2 9 - 10 3951 70
Gene Symbol: apc-17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063980.3 NP_496381.1 12 - 13 4952 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter transport
cell cycle
metaphase/anaphase transition of mitotic cell cycle
multicellular organism development
asymmetric cell division
polarity specification of anterior/posterior axis
protein ubiquitination
anaphase-promoting complex-dependent catabolic process
metaphase/anaphase transition of meiotic cell cycle
establishment of meiotic spindle localization
cell division
meiotic cell cycle
protein K11-linked ubiquitination
binding, bridging
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0