This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02438617_g1
See other MPZ-1 GE Assays ›
Gene Symbol:
mpz-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C52A11.4
Chromosome Location:
Chr.II: 10738635 - 10778126 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02438617_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mpz-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026867.4 NP_001022038.2 41 - 42 7133 60
NM_001083157.4 NP_001076626.2 35 - 36 6526 60
NM_001129127.4 NP_001122599.1 34 - 35 6472 60
NM_001129129.4 NP_001122601.1 39 - 40 6967 60
NM_001267333.3 NP_001254262.1 18 - 19 4147 60
NM_001267334.3 NP_001254263.1 11 - 12 2860 60
NM_001267335.3 NP_001254264.1 6 - 7 1330 60
NM_001377853.2 NP_001364655.1 42 - 43 7186 60
NM_001377854.2 NP_001364657.1 41 - 42 7111 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

protein binding
GTPase activating protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0