This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02438592_g1
See other MPZ-1 GE Assays ›
Gene Symbol:
mpz-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C52A11.4
Chromosome Location:
Chr.II: 10738635 - 10778126 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02438592_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mpz-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026867.4 NP_001022038.2 16 - 17 1730 79
NM_001026868.5 NP_001022039.2 16 - 17 1711 79
NM_001083157.4 NP_001076626.2 10 - 11 1123 79
NM_001129127.4 NP_001122599.1 9 - 10 1069 79
NM_001129129.4 NP_001122601.1 14 - 15 1552 79
NM_001377853.2 NP_001364655.1 17 - 18 1783 79
NM_001377854.2 NP_001364657.1 16 - 17 1696 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

protein binding
GTPase activating protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0