This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02438031_g1
See other PLC-3 GE Assays ›
Gene Symbol:
plc-3
Gene Name:
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma plc-3
Gene Aliases:
CELE_T01E8.3
Chromosome Location:
Chr.II: 10222877 - 10229440 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02438031_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plc-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063804.5 NP_496205.2 4 - 5 1106 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
signal transduction
phospholipid catabolic process
positive regulation of epithelial cell migration
lipid catabolic process
sleep
intracellular signal transduction
phosphatidylinositol phospholipase C activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0